Azathioprine
Synonym(s):
6-(1-Methyl-4-nitroimidazol-5-yl)thiopurine
Empirical Formula (Hill Notation):
C9H7N7O2S
CAS Number:
Molecular Weight:
277.26
Products
Shipping
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing