Skip to Content
Merck
Astilbin from Engelhardtia roxburghiana ≥98%

Astilbin from Engelhardtia roxburghiana

Astilbin from Engelhardtia roxburghiana ≥98%
Synonym(s):
Astilbin, Dihydroquercetin 3-rhamnoside, Taxifolin 3-rhamnoside
Empirical Formula (Hill Notation):
C21H22O11
CAS Number:
Molecular Weight:
450.39

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing