Skip to Content
Merck
Arsenobetaine purum p.a., ≥95.0% (NMR)

Arsenobetaine

Arsenobetaine purum p.a., ≥95.0% (NMR)
Synonym(s):
2-(Trimethylarsonio)acetate
Empirical Formula (Hill Notation):
C5H11AsO2
CAS Number:
Molecular Weight:
178.06

Products

Shipping
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing