Skip to Content
Merck
Arecoline hydrobromide VETRANAL®, analytical standard

Arecoline hydrobromide

Arecoline hydrobromide VETRANAL®, analytical standard
Synonym(s):
1-Methyl-1,2,5,6-tetrahydro-3-pyridinecarboxylic acid methyl ester hydrobromide, Arecaidine methyl ester hydrobromide, Methyl 1,2,5,6-tetrahydro-1-methyl-3-pyridinecarboxylate hydrobromide, Methyl 1-methyl-1,2,5,6-tetrahydronicotinate hydrobromide, Taeniolin
Empirical Formula (Hill Notation):
C8H13NO2 · HBr
CAS Number:
Molecular Weight:
236.11
EC Number:

Products

Shipping
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2