![Arcaine sulfate salt](/deepweb/assets/sigmaaldrich/product/structures/545/767/ee1a2e56-e74f-46af-8b77-e4031a1b6002/640/ee1a2e56-e74f-46af-8b77-e4031a1b6002.png)
Arcaine sulfate salt
![Arcaine sulfate salt](/deepweb/assets/sigmaaldrich/product/structures/545/767/ee1a2e56-e74f-46af-8b77-e4031a1b6002/640/ee1a2e56-e74f-46af-8b77-e4031a1b6002.png)
Synonym(s):
1,4-Diguanidinobutane sulfate salt
Empirical Formula (Hill Notation):
C6H16N6 · H2O4S
CAS Number:
Molecular Weight:
270.31
Products
Shipping
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing