Skip to Content
Merck
Arachidonic acid 1.0 mg/mL in ethanol, certified reference material, Cerilliant®

Arachidonic acid

Arachidonic acid 1.0 mg/mL in ethanol, certified reference material, Cerilliant®
Synonym(s):
cis,cis,cis,cis-5,8,11,14-Eicosatetraenoic acid, Eicosa-5Z,8Z,11Z,14Z-tetraenoic acid, Immunocytophyte, Arachidonic Acid
Linear Formula:
CH3(CH2)4(CH=CHCH2)4CH2CH2CO2H
CAS Number:
Molecular Weight:
304.47

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Research Area
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique