
Apocholic acid

Synonym(s):
8(14),5β-Cholenoic acid-3α,12α-diol
Empirical Formula (Hill Notation):
C24H38O4
CAS Number:
Molecular Weight:
390.56
Products
Product Category
Formula Weight
Manufacturer
Physical Form
Purity
Package Size
Showing 1-1 of 1
Compare
Product Number
Description
Pricing