Skip to Content
Merck
Anti-Vascular Endothelial Growth Factor Receptor-2 (KDR)−Biotin Conjugate antibody, Mouse monoclonal clone 260.4, KDR-2, purified from hybridoma cell culture

Anti-Vascular Endothelial Growth Factor Receptor-2 (KDR)−Biotin Conjugate antibody, Mouse monoclonal

Anti-Vascular Endothelial Growth Factor Receptor-2 (KDR)−Biotin Conjugate antibody, Mouse monoclonal clone 260.4, KDR-2, purified from hybridoma cell culture
Synonym(s):
Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-2 antibody produced in mouse, Anti-KDR, Anti-VEGFR-2

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique