Anti-SH3KBP1 antibody produced in rabbit
Synonym(s):
Anti-CD2-binding protein 3 antibody produced in rabbit, Anti-CD2BP3 antibody produced in rabbit, Anti-Cbl-interacting protein of 85 kDa antibody produced in rabbit, Anti-HSB-1 antibody produced in rabbit, Anti-Human Src-family kinase-binding protein 1 antibody produced in rabbit, Anti-SH3 domain-containing kinase-binding protein 1 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing