Skip to Content
Merck
Anti-c-Myc−Biotin antibody, Mouse monoclonal clone 9E10, purified from hybridoma cell culture

Anti-c-Myc−Biotin antibody, Mouse monoclonal

Anti-c-Myc−Biotin antibody, Mouse monoclonal clone 9E10, purified from hybridoma cell culture
Synonym(s):
Anti-c-Myc

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1