
Anti-CDC5L antibody produced in rabbit

Synonym(s):
Anti-Cdc5-like protein antibody produced in rabbit, Anti-Cell division cycle 5-like protein antibody produced in rabbit, Anti-Pombe cdc5-related protein antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing