Skip to Content
Merck
α -Nitrotoluene 95%

α-Nitrotoluene

α -Nitrotoluene 95%
Synonym(s):
Nitromethylbenzene, Phenylnitromethane
Linear Formula:
C6H5CH2NO2
CAS Number:
Molecular Weight:
137.14

Products

Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing