Skip to Content
Merck
Ammonium nitrate-15N 60 atom % 15N

Ammonium nitrate-15N

Ammonium nitrate-15N 60 atom % 15N
Synonym(s):
15N ammonium salt, 15N labeled ammonium nitrate
Linear Formula:
NH415NO3
CAS Number:
Molecular Weight:
81.04

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Application
Markush Class
Markush Group
Showing 1-4 of 4
Compare
Product Number
Description
Pricing