Skip to Content
Merck
Amitriptyline hydrochloride British Pharmacopoeia (BP) Reference Standard

Amitriptyline hydrochloride

Amitriptyline hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Amitriptyline hydrochloride
Empirical Formula (Hill Notation):
C20H23N · HCl
CAS Number:
Molecular Weight:
313.86

Products

Shipping
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique