Skip to Content
Merck
Aminobenzoate Potassium pharmaceutical secondary standard, certified reference material

Aminobenzoate potassium

Aminobenzoate Potassium pharmaceutical secondary standard, certified reference material
Synonym(s):
4-Aminobenzoic acid potassium salt, PABA, Potassium 4-aminobenzoate
Linear Formula:
C7H6NO2K
CAS Number:
Molecular Weight:
175.23

Products

Shipping
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Special Grade