
ALX 5407 hydrochloride

Synonym(s):
ALX-5407 hydrochloride, N-[3-(4′-Fluorophenyl)-3-(4′-phenylphenoxy)propyl]sarcosine hydrochloride, NFPS hydrochloride
Empirical Formula (Hill Notation):
C24H24NO3F·HCl
CAS Number:
Molecular Weight:
429.91
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1