Skip to Content
Merck
Aluminum phosphate monobasic purum, ≥95.0% (gravimetric)

Aluminum phosphate monobasic

Aluminum phosphate monobasic purum, ≥95.0% (gravimetric)
Synonym(s):
Aluminum dihydrogen phosphate, prim-Aluminum phosphate
Linear Formula:
Al(H2PO4)3
CAS Number:
Molecular Weight:
317.94
EC Number:

Products

Shipping
Formula Weight
Physical Form
Purity
Quality Segment
Reagent Type
Core
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing