Skip to Content
Merck
Alendronate sodium trihydrate ≥97% (NMR), powder

Alendronate sodium trihydrate

Alendronate sodium trihydrate ≥97% (NMR), powder
Synonym(s):
4-amino-1-hydroxy-1-phosphonobutyl phosphonic acid, monosodium, MK-217
Empirical Formula (Hill Notation):
C4H12NaNO7P2 · 3 H2O
CAS Number:
Molecular Weight:
325.12

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing