![Alendronate Sodium Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/245/006/45632b44-67b2-4f1b-8504-8e301a249294/640/45632b44-67b2-4f1b-8504-8e301a249294.png)
Alendronate sodium
![Alendronate Sodium Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/245/006/45632b44-67b2-4f1b-8504-8e301a249294/640/45632b44-67b2-4f1b-8504-8e301a249294.png)
Synonym(s):
Alendronate sodium trihydrate, 4-amino-1-hydroxy-1-phosphonobutyl phosphonic acid, monosodium, MK-217
Empirical Formula (Hill Notation):
C4H12NaNO7P2 · 3 H2O
CAS Number:
Molecular Weight:
325.12
Products
Shipping
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Technique
Showing 1-2 of 2