AIDA
Synonym(s):
1-Amino-2,3-dihydro-1H-indene-1,5-dicarboxylic acid, 1-Aminoindan-1, 5-dicarboxylic acid, UPF 523
Empirical Formula (Hill Notation):
C11H11NO4
CAS Number:
Molecular Weight:
221.21
Products
Shipping
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing