![Adrenaline acid tartrate British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/388/173/778986a2-927b-4ae1-8696-1b20cc05061f/640/778986a2-927b-4ae1-8696-1b20cc05061f.png)
Adrenaline acid tartrate
![Adrenaline acid tartrate British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/388/173/778986a2-927b-4ae1-8696-1b20cc05061f/640/778986a2-927b-4ae1-8696-1b20cc05061f.png)
Synonym(s):
(−)-Epinephrine (+)-bitartrate salt, L-3,4-Dihydroxy-α-(methylaminomethyl)benzyl alcohol D-hydrogen bitartrate salt, L-Adrenaline (+)-bitartrate salt, L-Adrenaline D-hydrogentartrate, L-Epinephrine D-hydrogentartrate
Empirical Formula (Hill Notation):
C9H13NO3 · C4H6O6
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing