Adapalene Related Compound C
Synonym(s):
o-Adamantylanisole, 1-(2-Methoxyphenyl)-tricyclo[3.3.1.13,7]decane, 2-(Adamant-1-yl)methoxybenzene
Empirical Formula (Hill Notation):
C17H22O
CAS Number:
Molecular Weight:
242.36
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2