Skip to Content
Merck
α-Cyclodextrin purum, ≥98.0% (HPLC)

α-Cyclodextrin

α-Cyclodextrin purum, ≥98.0% (HPLC)
Synonym(s):
α-Schardinger dextrin, Cyclohexaamylose, Cyclomaltohexaose
Empirical Formula (Hill Notation):
C36H60O30
CAS Number:
Molecular Weight:
972.84
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Sterility
Technique
Showing 1-5 of 5