Skip to Content
DE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Activin A human
Synonym(s):
Activin beta-A, FRP, Inhibin beta-1
Products
Product Category
growth factors (3)
peptides (1)
Brand
Sigma-Aldrich (3)
Biological Source
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
SRP3003
Activin A human,
recombinant, expressed in
E. coli
, ≥97% (SDS-PAGE), ≥97% (HPLC), suitable for cell culture
Expand
View Pricing
A4941
Activin A human,
≥95% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, lyophilized powder, suitable for cell culture
Expand
View Pricing
H4666
Activin A human,
recombinant, expressed in HEK 293 cells, HumanKine
®
, suitable for cell culture
Expand
View Pricing