Acrinathrin
Synonym(s):
(S)-Cyano(3-phenoxyphenyl)methyl (Z)-(1R,3S)-2,2-dimethyl[2-(2,2,2-trifluoro-1-trifluoromethylethoxycarbonyl)vinyl]-cyclopropanecarboxylate
Empirical Formula (Hill Notation):
C26H21F6NO5
CAS Number:
Molecular Weight:
541.44
Products
Shipping
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2