Skip to Content
Merck
Acetyl fentanyl-13C6 solution 50 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Acetyl fentanyl-13C6 solution

Acetyl fentanyl-13C6 solution 50 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®
Empirical Formula (Hill Notation):
13C6C15H26N2O
Molecular Weight:
328.40

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique