Skip to Content
Merck
Acemetacin impurity A European Pharmacopoeia (EP) Reference Standard

Acemetacin impurity A

Acemetacin impurity A European Pharmacopoeia (EP) Reference Standard
Synonym(s):
4-Chlorobenzoic acid
Linear Formula:
ClC6H4CO2H
CAS Number:
Molecular Weight:
156.57

Products

Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing