α-Asarone
Synonym(s):
trans-1,2,4-Trimethoxy-5-(1-propenyl)benzene, trans-1-Propenyl-2,4,5-trimethoxybenzene, trans-Asarone, Isoasaron
Linear Formula:
(CH3O)3C6H2CH=CHCH3
CAS Number:
Molecular Weight:
208.25
Products
Shipping
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing