![8-Azaguanine ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/333/565/dc1980d5-e186-4aa1-a667-54482d5e38f3/640/dc1980d5-e186-4aa1-a667-54482d5e38f3.png)
8-Azaguanine
![8-Azaguanine ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/333/565/dc1980d5-e186-4aa1-a667-54482d5e38f3/640/dc1980d5-e186-4aa1-a667-54482d5e38f3.png)
Synonym(s):
2-Amino-6-hydroxy-8-azapurine, 2-Amino-6-oxy-8-azapurine, Guanazolo
Empirical Formula (Hill Notation):
C4H4N6O
CAS Number:
Molecular Weight:
152.11
Products
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing