![8arm-PEG20K-COOH, tripentaerythritol core average Mn 20,000](/deepweb/assets/sigmaaldrich/product/structures/833/300/18f98028-c071-43d8-a4d4-cc5c71be1cee/640/18f98028-c071-43d8-a4d4-cc5c71be1cee.png)
8arm-PEG20K-COOH, tripentaerythritol core
![8arm-PEG20K-COOH, tripentaerythritol core average Mn 20,000](/deepweb/assets/sigmaaldrich/product/structures/833/300/18f98028-c071-43d8-a4d4-cc5c71be1cee/640/18f98028-c071-43d8-a4d4-cc5c71be1cee.png)
Synonym(s):
8arm-PEG-COOH, tripentaerythritol core
Linear Formula:
R(O(CH2CH2O)nCH2COOH)8
Products
Brand
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing