![Rifampicin ≥95% (HPLC), powder or crystals](/deepweb/assets/sigmaaldrich/product/structures/384/200/dc7204ed-b3e1-45f7-b463-91188e2328ab/640/dc7204ed-b3e1-45f7-b463-91188e2328ab.png)
Rifampicin
![Rifampicin ≥95% (HPLC), powder or crystals](/deepweb/assets/sigmaaldrich/product/structures/384/200/dc7204ed-b3e1-45f7-b463-91188e2328ab/640/dc7204ed-b3e1-45f7-b463-91188e2328ab.png)
Synonym(s):
3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifampin, Rifamycin AMP, Rifampicin
Empirical Formula (Hill Notation):
C43H58N4O12
CAS Number:
Molecular Weight:
822.94
Products
Product Category
Brand
Color
Formula Weight
Physical Form
pK Value
Product Line
Purity
Activity Spectrum
Quality Segment
Application
Mode of Action
Technique
Showing 1-3 of 3