
6-Methoxy-2-naphthol

Synonym(s):
2-Hydroxy-6-methoxynaphthalene, 6-Methoxynaphthalen-2-ol
Empirical Formula (Hill Notation):
C11H10O2
CAS Number:
Molecular Weight:
174.20
Products
Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1