Skip to Content
Merck
6-Chloro-3-methyluracil ≥98%

6-Chloro-3-methyluracil

6-Chloro-3-methyluracil ≥98%
Synonym(s):
6-Chloro-3-methylpyrimidine-2,4(1H,3H)-dione
Empirical Formula (Hill Notation):
C5H5ClN2O2
CAS Number:
Molecular Weight:
160.56

Products

Shipping
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing