Skip to Content
Merck
5-Methylcytosine hydrochloride ≥99%

5-Methylcytosine hydrochloride

5-Methylcytosine hydrochloride ≥99%
Synonym(s):
4-Amino-2-hydroxy-5-methylpyrimidine
Empirical Formula (Hill Notation):
C5H7N3O · HCl
CAS Number:
Molecular Weight:
161.59
EC Number:

Products

Shipping
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing