
5-Hydroxymethylcytosine

Empirical Formula (Hill Notation):
C5H7N3O2
CAS Number:
Molecular Weight:
141.13
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Physical Form
Research Area
Quality Segment
Application
Showing 1-1 of 1