Skip to Content
Merck
5-Ethynylpyrimidine 95%

5-Ethynylpyrimidine

5-Ethynylpyrimidine 95%
Empirical Formula (Hill Notation):
C6H4N2
CAS Number:
Molecular Weight:
104.11

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing