Skip to Content
Merck
5-Ethyl-3,7-dioxa-1-azabicyclo[3.3.0]octane analytical standard

5-Ethyl-3,7-dioxa-1-azabicyclo[3.3.0]octane

5-Ethyl-3,7-dioxa-1-azabicyclo[3.3.0]octane analytical standard
Synonym(s):
Dihydro-7a-ethyloxazolo[3,4-c]oxazole
Empirical Formula (Hill Notation):
C7H13NO2
CAS Number:
Molecular Weight:
143.18

Products

Shipping
Brand
Boiling Point (°C)
Formula Weight
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing