Skip to Content
Merck
5-Carboxyfluorescein BioReagent, suitable for fluorescence, ≥95% (HPLC)

5-Carboxyfluorescein

5-Carboxyfluorescein BioReagent, suitable for fluorescence, ≥95% (HPLC)
Synonym(s):
5-FAM, 5-Carboxyfluorescein
Empirical Formula (Hill Notation):
C21H12O7
CAS Number:
Molecular Weight:
376.32

Products

Shipping
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Reagent Type
Showing 1-2 of 2