
5-Bromoindoxyl acetate

Synonym(s):
3-Acetoxy-5-bromoindole, O-Acetyl-5-bromoindoxyl
Empirical Formula (Hill Notation):
C10H8BrNO2
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing