Skip to Content
Merck
5-Bromo-3-nitropyridine-2-carbonitrile 95%

5-Bromo-3-nitropyridine-2-carbonitrile

5-Bromo-3-nitropyridine-2-carbonitrile 95%
Empirical Formula (Hill Notation):
C6H2BrN3O2
CAS Number:
Molecular Weight:
228.00

Products

Shipping
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing