Skip to Content
Merck
5-Amino-2-nitrobenzoic acid 97%

5-Amino-2-nitrobenzoic acid

5-Amino-2-nitrobenzoic acid 97%
Synonym(s):
2-Nitro-5-aminobenzoic acid, 3-Carboxy-4-nitroaniline, NSC 74455
Linear Formula:
H2NC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
182.13
EC Number:

Products

Shipping
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing