Skip to Content
Merck
5,6-Dimethyl-2-thiouracil 97%

5,6-Dimethyl-2-thiouracil

5,6-Dimethyl-2-thiouracil 97%
Empirical Formula (Hill Notation):
C6H8N2OS
CAS Number:
Molecular Weight:
156.21

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing