4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt
Synonym(s):
Sulfo-SMCC
Empirical Formula (Hill Notation):
C16H17N2NaO9S
CAS Number:
Molecular Weight:
436.37
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Application
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing