Skip to Content
Merck
4-Methyl-5-thiazoleethanol analytical standard

4-Methyl-5-thiazoleethanol

4-Methyl-5-thiazoleethanol analytical standard
Synonym(s):
5-(2-Hydroxyethyl)-4-methylthiazole, Sulfurol
Empirical Formula (Hill Notation):
C6H9NOS
CAS Number:
Molecular Weight:
143.21
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing