4-Methyl-4-mercaptopentan-2-one
Empirical Formula (Hill Notation):
C6H12OS
CAS Number:
Molecular Weight:
132.22
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing