![4-Hydroxybenzophenone 98%](/deepweb/assets/sigmaaldrich/product/structures/241/416/cc0080f8-d79a-45f9-9257-efbf25953eeb/640/cc0080f8-d79a-45f9-9257-efbf25953eeb.png)
4-Hydroxybenzophenone
![4-Hydroxybenzophenone 98%](/deepweb/assets/sigmaaldrich/product/structures/241/416/cc0080f8-d79a-45f9-9257-efbf25953eeb/640/cc0080f8-d79a-45f9-9257-efbf25953eeb.png)
Synonym(s):
4-Hydroxybenzophenone
Linear Formula:
4-(OH)C6H4COC6H5
CAS Number:
Molecular Weight:
198.22
Products
Shipping
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing