Skip to Content
Merck
4-Formylphenylboronic acid ≥95.0%

4-Formylphenylboronic acid

4-Formylphenylboronic acid ≥95.0%
Synonym(s):
4-(Dihydroxyboryl)benzaldehyde, 4-Boronobenzaldehyde, 4-Formylbenzeneboronic acid, p-Formylbenzeneboronic acid, p-Formylphenylboronic acid
Linear Formula:
HCOC6H4B(OH)2
CAS Number:
Molecular Weight:
149.94

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing