Skip to Content
Merck
4-Chloro-1-butanol technical grade, ~85%

4-Chloro-1-butanol

4-Chloro-1-butanol technical grade, ~85%
Synonym(s):
Tetramethylene chlorohydrin
Linear Formula:
HO(CH2)4Cl
CAS Number:
Molecular Weight:
108.57

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing