Skip to Content
Merck
4-(Butylamino)benzoic acid 97%

4-(Butylamino)benzoic acid

4-(Butylamino)benzoic acid 97%
Linear Formula:
CH3(CH2)3NHC6H4CO2H
CAS Number:
Molecular Weight:
193.24

Products

Shipping
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing