Skip to Content
Merck
4-Bromothiophene-2-acetic acid 97%

4-Bromothiophene-2-acetic acid

4-Bromothiophene-2-acetic acid 97%
Empirical Formula (Hill Notation):
C6H5BrO2S
CAS Number:
Molecular Weight:
221.07

Products

Shipping
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing